General-purpose masking tape 201E
Supplier: 3M
201E-18MM 201E-24MM 201E-30MM 201E-36MM 201E-48MM
MMMA201E-18MMEA 3 EUR
MMMA201E-18MM MMMA201E-24MM MMMA201E-30MM MMMA201E-36MM MMMA201E-48MM
General-purpose masking tape 201E
Tapes
This is a premium-quality masking tape used for short-term paint masking, fixing, bundling, holding, sealing, and other applications up to a temperature of 80 °C for one hour.
  • Good instant adhesion and holding power that resists lifting or curling
  • Operating temperature range up to 80 °C for one hour
  • Reduces paint build-up and provides good paint lines, helps resist paint bleed through
  • Designed for indoor use
This tape is designed for general use in industrial applications with a slight need for temperature and water resistance, such as short-termMore Product Information