Powered and supplied air breathing tubes
Supplier: 3M
S222
MMMAS222EA 395 EUR
MMMAS222
Powered and supplied air breathing tubes
Respirators Respiratory Systems
Breathing Tube S-222 is a replacement breathing tube for use with 3M™ Versaflo™ Supplied Air System S-200+.
  • Spare or replacement part
  • Protective breathing tube covers available to lengthen service life of breathing tubes
Prolong the service life of your 3M™ Versaflo™ Supplied Air System S-200+ with the Breathing Tube, S-222. This spare breathing tube is specially designed for use with S-200+ systems and is CE approved.