Tegaderm™ HP+Pad Film Dressing with Non-Adherent Pad
Supplier: 3M MEDICAL
8582 8584
MMME8582EA 163 EUR
MMME8582 MMME8584
Tegaderm™ HP+Pad Film Dressing with Non-Adherent Pad
First Aid Supplies Wound Dressings
The Tegaderm™ +Pad Film Dressing is a sterile surgical dressing with a central absorbent and non-adherent pad that helps in moist wound healing and painless dressing removal. This Tegaderm dressing is breathable, waterproof, bacterial and viral barrier thus protecting against infections and is easy to maintain.
  • Bacterial and Viral barrier to prevent surgical site infections
  • Waterproof barrier which allows patients to bathe or shower without removing dressing
  • Unique, absorbent, non-adherent pad won`t stick to the wound, allowing moist wound healing with