Swab-sampler with letheen broth
Supplier: 3M Food Safety
RS96010LET RS9601LET RS9604LET
MMMPRS96010LETEA 143 EUR
MMMPRS96010LET MMMPRS9601LET MMMPRS9604LET
Swab-sampler with letheen broth
Samplers Solid Samplers
3M™ Swab-sampler with Letheen broth is a convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Letheen broth vial is ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Sample collection is easy with theMore Product Information