2-Amino-4-methylpyridine

Supplier: Molekula

Synonyms: Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine

11240887-100G
MOLE11240887-100GEA 0 EUR
MOLE11240887-100G
2-Amino-4-methylpyridine
2-Amino-4-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 230.9 °C (1013 hPa)
Melting Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006321
CAS Number: 695-34-1
EINECS: 211-780-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,00465

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR