2-Naphthylacetic acid

Supplier: Molekula

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

12224581-5G
MOLE12224581-5GEA 0 EUR
MOLE12224581-5G
2-Naphthylacetic acid
2-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352
Melting Pt: 139…142 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR