Propidium iodide

Supplier: Molekula
Warning

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide

14137305-100MG
MOLE14137305-100MGEA 358 EUR
MOLE14137305-100MG
Propidium iodide
Propidium iodide
Formula: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
MDL Number: MFCD00011921
CAS Number: 25535-16-4
EINECS: 247-081-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR