Salicylamide

Supplier: Molekula
Warning

Synonyms: 2-Hydroxybenzamide

19861860-1KG
MOLE19861860-1KGEA 0 EUR
MOLE19861860-1KG
Salicylamide
Salicylamide
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 318
Melting Pt: 137
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00007978
CAS Number: 65-45-2
EINECS: 200-609-3
Merck Index: 12,08480

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR