Benzalkonium chloride 50% aqueous solution

Supplier: Molekula
Danger

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

29089235-500ML
MOLE29089235-500MLEA 0 EUR
MOLE29089235-500ML
Benzalkonium chloride 50% aqueous solution
Benzalkonium chloride
Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR