1-Naphthylacetic acid

Supplier: Molekula
Warning

Synonyms: Planofix, 1-Naphthaleneacetic acid

35143629-100G
MOLE35143629-100GEA 144 EUR
MOLE35143629-100G
1-Naphthylacetic acid
1-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352
Melting Pt: 129…131 °C
Density: ∼1.263 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004046
CAS Number: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR