4-Chloro-α,α,α-trifluorotoluene

Supplier: Molekula
Danger

Synonyms: 1-Chloro-4-(trifluoromethyl)benzene, 4-Chlorobenzotrifluoride

36946440-500G
MOLE36946440-500GEA 0 EUR
MOLE36946440-500G
4-Chloro-α,α,α-trifluorotoluene
4-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 136 °C (1013 hPa)
Melting Pt: –36 °C
Density: 1.346 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000627
CAS Number: 98-56-6
EINECS: 202-681-1
UN: 2234
ADR: 3,III
Merck Index: 13,02145

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR