2-Thiophenemethanol

Supplier: Molekula

Synonyms: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol

40682099-25G
MOLE40682099-25GEA 0 EUR
MOLE40682099-25G
2-Thiophenemethanol
2-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.206 g/cm³ (20 °C)
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR