ω-Bromo-3,4-dichloroacetophenone

Supplier: Molekula

Synonyms: 2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one, 2-Bromo-1-(3,4-dichlorophenyl)ethanone, 2-Bromo-3',4'-dichloroacetophenone, 3,4-Dichlorophenacyl bromide, α-Bromo-3,4-dichloroacetophenone

44437650-5G
MOLE44437650-5GEA 68 EUR
MOLE44437650-5G
ω-Bromo-3,4-dichloroacetophenone
ω-Bromo-3,4-dichloroacetophenone
Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00051581
CAS Number: 2632-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR