Calcium-D gluconate

Supplier: Molekula

Synonyms: Calcium Gluconate, Calcium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconate Calcium, D-Gluconic acid calcium salt

51659757-100G
MOLE51659757-100GEA 0 EUR
MOLE51659757-100G
Calcium-D gluconate
Calcium D-gluconate
Formula: HOH₂C(CH(OH))₄COOCa
MW: 430.38 g/mol
Melting Pt: 178
MDL Number: MFCD00064209
CAS Number: 299-28-5
EINECS: 206-075-8
Merck Index: 13,01671

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR