2,4-Dimethoxyaniline

Supplier: Molekula

55247174-100G
MOLE55247174-100GEA 0 EUR
MOLE55247174-100G
2,4-Dimethoxyaniline
2,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 39 °C
Flash Pt: >130 °C
Storage Temperature: Ambient
MDL Number: MFCD00008371
CAS Number: 2735-04-8
EINECS: 220-355-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR