Sodium alginate

Supplier: Molekula

Synonyms: Alginic acid sodium salt, Algin

60689063-1KG
MOLE60689063-1KGEA 0 EUR
MOLE60689063-1KG
Sodium alginate
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR