Bronopol

Supplier: Molekula
Danger

Synonyms: 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol

65898637-100G
MOLE65898637-100GEA 138 EUR
MOLE65898637-100G
Bronopol
Bronopol
Formula: HOCH₂BrC(NO₂)CH₂OH
MW: 199.99 g/mol
Boiling Pt: 388
Melting Pt: 130…133 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007390
CAS Number: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: 4.1 III
Merck Index: 13,01432

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR