6-Benzylaminopurine

Supplier: Molekula

Synonyms: 6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine

M48043715
MOLEM48043715EA 0 EUR
MOLEM48043715
6-Benzylaminopurine
6-Benzylaminopurine
Formula: C₁₂H₁₁N₅
MW: 225.25 g/mol
Melting Pt: 230…234 °C
MDL Number: MFCD00005572
CAS Number: 1214-39-7
EINECS: 214-927-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR