Ammonium benzoate

Supplier: Molekula
Warning

Synonyms: Benzoic acid ammonium salt

M62761848
MOLEM62761848EA 0 EUR
MOLEM62761848
Ammonium benzoate
Ammonium benzoate
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 197.8
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013069
CAS Number: 1863-63-4
EINECS: 217-468-9
Merck Index: 12,00521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR