1-(4-Nitrophenyl)piperazine

Supplier: Molekula

M65396606
MOLEM65396606EA 0 EUR
MOLEM65396606
1-(4-Nitrophenyl)piperazine
1-(4-Nitrophenyl)piperazine
Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Melting Pt: 136.85 °C
MDL Number: MFCD00005961
CAS Number: 6269-89-2
EINECS: 228-443-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR