1-Bromo-4-butylbenzene

Supplier: Molekula

Synonyms: (4-Bromophenyl)butane

M86441856
MOLEM86441856EA 0 EUR
MOLEM86441856
1-Bromo-4-butylbenzene
1-Bromo-4-butylbenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –26 °C
Density: 1,2347 g/cm³ (20 °C)
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR