3,3',5,5'-Tetramethylbenzidine (TMB)

Supplier: Molekula

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

M88604857
MOLEM88604857EA 0 EUR
MOLEM88604857
3,3',5,5'-Tetramethylbenzidine (TMB)
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR