Safety storage cabinets, type 90, S-PHOENIX-90
Supplier: asecos
30313001 30313002 30314001 30314002 30233001 30233002 30118001 30118002 30234001 30234002 30207001 30207002
MSGA30313001EA 2737 EUR
MSGA30313001 MSGA30313002 MSGA30314001 MSGA30314002 MSGA30233001 MSGA30233002 MSGA30118001 MSGA30118002 MSGA30234001 MSGA30234002 MSGA30207001 MSGA30207002
Safety storage cabinets, type 90, S-PHOENIX-90
Furniture Storage Cabinets
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 min).
  • Space saving folding doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Door open arrest system (optional), convenient and safe removal or storage of containers
  • Doors lockable with cylinder locking (integration in a locking system possible)
  • Adjusting aids to compensate for uneven floor
  • Integrated air ducts ready for connection (NW 75) to a technical exhaust system
Certifikace: EN 14727 conformity (laboratory furniture).More Product Information