Bottles, wide neck, with screw cap
Supplier: MURRAY S
CO120UC72
MURRCO120UC72EA 139 EUR
MURRCO120UC72
Bottles, wide neck, with screw cap
Bottles Glass Bottles
Clear glass, with black Bakelite closure.
  • Screw closure with paraffin wax lining
  • Good chemical resistance
  • Extra wide opening for easy filling and emptying
Can be used in all applications involving paste-like or powdery materials and liquids.