Dropping bottles, LDPE, Nalgene®
Supplier: Thermo Fisher Scientific
312751-9025 312750-9050 312750-9125 312751-9125 312750-9025 312751-9050
215-2350EA 1090 EUR
215-2350 NALG312750-9050 NALG312750-9125 NALG312751-9125 NALG312750-9025 NALG312751-9050
Dropping bottles, LDPE, Nalgene®
Bottles Dropping Bottles
Dropper bottles made up of natural/white low density polyethylene.
  • Round shape
  • Drop size: 40/50 μl
  • Leakproof
Certifikace: Compliance with ISO 13485:2003; 21CFR177.1520 and USP Class VI.
Informace pro objednávání: Controlled-drop dispensing tip and coloured closures are available seperately.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement