Septum caps, Nalgene®
Supplier: Thermo Fisher Scientific
342178-0384 342178-0240
NALG342178-0384EA 1090 EUR
NALG342178-0384 NALG342178-0240
Septum caps, Nalgene®
Closures Bottle Closures
HDPE with silicone/PTFE septum.
  • Sterile 10⁻⁶ SAL
  • No adhesives used to attach septum to high density polyethylene closure
  • Non-cytotoxic and non-pyrogenic
  • Dual septum material for improved gas barrier
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement