Counting chambers, C-Chip
Supplier: NANOENTEK
DHC-B01 DHC-B02 DHC-F01 DHC-M01 DHC-N01 DHC-N04
631-1094EA 255 EUR
631-1094 631-1095 631-1096 631-1097 631-1098 NANODHC-N04
Counting chambers, C-Chip
Slides Counting Chambers
Quartz grade optical plastic counting chambers applicable in blood analysis (hematology).
  • Guaranteed repeatability and reliability, eliminate the need for washing and reuse
  • Disposable hemocytometers that do not need coverslips
  • Chip to reduce exposure to infectious samples and contamination
Suitable for cell counting and culture, concentration measurement and viability test, in bacteria and fungal spore counting, IVF, IUI tasks of sperm counting.