Tubing Connectors, Luer-Lock
Supplier: NEOLAB
2-1889
NEOL2-1889EA 33.5 EUR
NEOL2-1889
Tubing Connectors, Luer-Lock
Fittings and Connectors
Tube Luer-connector with olive and Luer-Lock cone.
  • Made of natural coloured PP
  • Slightly transparent
  • Autoclavable 121 °C
Excellent quality with constant inner diameter, causes uniform flow rates and tight connection. The olives are worked on the outside of the sealing cone in such a way that solid tubing can also be attached. They expand the hose up to 1,5 times its inner diameter.
Balení: In a bag of 10 pieces.