Bulb pipettes, one mark, Class B
Supplier: NEUBERT VOLUME GLASSWAERE
0272-30-200
NEUB0272-30-200EA 88.7 EUR
NEUB0272-30-200
Bulb pipettes, one mark, Class B
Pipettes Bulb Pipettes
Glass.
  • Calibrated to deliver 'EX'
  • Full delivery
  • Blue graduations
Certifikace: DIN 12691