Apparatus with flange for the determination of Arsenic
Supplier: NEUBERT VOLUME GLASSWAERE
23900041002920
NEUB23900041002920EA 544 EUR
NEUB23900041002920
Apparatus with flange for the determination of Arsenic
Distillation Components Distillation Sets
Borosilicate glass 3,3.
Apparatus for the estimation of Arsenic. Complete including two springs, erlenmeyer flasks 100 ml, graduated, ST 29/32, tube with ST 29/32 and flange.
Certifikace: DAB 10 (Pharmeuropa 28.2 ; Ph.Eur 2.4.2 Method A )