PIG® spill response bucket kit
Supplier: NEW PIG
64CRSC KIT213
NEWP64CRSCEA 125 EUR
NEWP64CRSC 121-0556
PIG® spill response bucket kit
Spill Control Materials
Just grab and go for fast response to spills up to 13,8 litres.
  • Container is UN-rated for shipping waste (UN1H2/Y30/S)
  • PIG socks stop spreading spills; PIG mat absorbs quickly
  • Prepacked container provides easy access to contents
  • Bold Spill Kit lettering for quick identification
  • Temporary disposal bags with ties help make cleanup easier
Tamperproof seals help prevent pilfering of spill response supplies.
Upozornení: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG®More Product Information