Absorbent mats, oil-only, 4 in 1®, PIG®
Supplier: NEW PIG
MAT435 MAT484
NEWPMAT435EA 170.75 EUR
NEWPMAT435 NEWPMAT484
Absorbent mats, oil-only, 4 in 1®, PIG®
Spill Control Materials
Highly absorbent mats are made from a eight-layer 100% PP medium-weight construction and function as pads, rolls, socks or wipes. They absorb and retain a variety of leaks, drips, and spills of oils and fuels, except water.
  • Scuff-resistant, tear-resistant, even under heavy foot traffic or abrasive conditions
  • Flame-resistant - material melts when exposed to high heat
  • Dimple pattern wicks of liquid throughout mat for faster and easier clean-up
  • Rolls are perforated every 250 mm