PIG® absorbent pillows
Supplier: NEW PIG
PIL201 PIL204 PIL205
121-0539EA 127 EUR
121-0539 NEWPPIL204 NEWPPIL205
PIG® absorbent pillows
Spill Control Materials
Add extra absorbency to your spill cleanup efforts with high-capacity PIG Pillows. Also great for steady leaks and drips.
  • Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Cellulose filler is highly absorbent for long-term use or big spill cleanup
  • Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response
  • Absorbs and retains oils, coolants, solvents and water;