Cleanroom sponge wipes, sterile, BioClean Isorba™

Supplier: Nitritex
BISW-8
NITRBISW-8EA 304 EUR
NITRBISW-8
Cleanroom sponge wipes, sterile, BioClean Isorba™
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
This sterile sponge wipe is mande of non-abrasive polyurethane foam. It is suitable for use on all surfaces.

  • Low particulate generation
  • Excellent absorption properties, strength and durability
  • Sterilised by gamma irradiation

Thickness: 3 mm
Density: 25 kg/m

Balení: Two sponge wipes per double bag, 25 double bags per outer bag; two outer bags per carton.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR