Lysates, mammalian GenLysate™, human
Supplier: G Bioscienes
NLH-01 NLH-02 NLH-03 NLH-04 NLH-05 NLH-06 NLH-08 NLH-09 NLH-11 NLH-12 NLH-13 NLH-14 NLH-15 NLH-16 NLH-17 NLH-18 NLH-19 NLH-20 NLH-21 NLH-22 SLH-021 SLH-0210 SLH-0211 SLH-0212 SLH-0213 SLH-023 SLH-024 SLH-025 SLH-026 SLH-027 SLH-028 SLH-029 SLH-081 SLH-082 SLH-087 SLH-088 SLH-089 SLH-090 TLH-01 TLH-03 TLH-06 TLH-07 TLH-09 TLH-11 TLH-12 TLH-13 TLH-14 TLH-15 TLH-16 TLH-17 TLH-18 TLH-19 TLH-20
NLH-01EA 308 EUR
NLH-01 NLH-02 NLH-03 NLH-04 NLH-05 NLH-06 NLH-08 NLH-09 NLH-11 NLH-12 NLH-13 NLH-14 NLH-15 NLH-16 NLH-17 NLH-18 NLH-19 NLH-20 NLH-21 NLH-22 SLH-021 SLH-0210 SLH-0211 SLH-0212 SLH-0213 SLH-023 SLH-024 SLH-025 SLH-026 SLH-027 SLH-028 SLH-029 SLH-081 SLH-082 SLH-087 SLH-088 SLH-089 SLH-090 TLH-01 TLH-03 TLH-06 TLH-07 TLH-09 TLH-11 TLH-12 TLH-13 TLH-14 TLH-15 TLH-16 TLH-17 TLH-18 TLH-19 TLH-20
Lysates, mammalian GenLysate™, human
Cells
GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes. The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with ProteaseARREST™ protease inhibitor cocktail. This minimises proteolytic damage to the proteins.
GenLysate™ total protein lysates are offered as a 150 µg lyophilised protein per vial and, following reconstitution in DI water, are ready for use. GenLysate™ is also available in a wide variety of other species and tissues. Species include: Human (both normal tissue and tumour tissue lysates available), mouse, rat, rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.