Anti-Mouse Monoclonal Antibody [clone: CC320] (Alexa Fluor 647)

Supplier: Novus Biologicals
NB100-63026AF6
NOVUNB100-63026AF6EA 882 EUR
NOVUNB100-63026AF6
Anti-Mouse Monoclonal Antibody [clone: CC320] (Alexa Fluor 647)
Antibodies
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: CC320
Conjugation: Alexa Fluor 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR