ND OLLAS/DYKDDDDK Epitope Tag Kit

Supplier: Novus Biologicals

NBK1-19033
NOVUNBK1-19033EA 572 EUR
NOVUNBK1-19033
ND OLLAS/DYKDDDDK Epitope Tag Kit
Assays Immunological Assays

ND OLLAS/DYKDDDDK Epitope Tag Kit


Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Non-species specific.

Order Now

SPECIFICATIONS

Environmentally Preferable
Species reactivity Non-species specific

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR