Anti-HIST4H4 Mouse Monoclonal Antibody (DyLight® 550) [clone: 5E10-D8]

Supplier: Novus Biologicals
NBP1-30091R
NOVUNBP1-30091REA 679 EUR
NOVUNBP1-30091R
Anti-HIST4H4 Mouse Monoclonal Antibody (DyLight® 550) [clone: 5E10-D8]
Antibodies
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: monoclonal
Clone: 5E10-D8
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR