Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)

Supplier: Novus Biologicals

NBP2-68071
NOVUNBP2-68071EA 1060 EUR
NOVUNBP2-68071
Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)
Assays ELISAs

This kit recognizes Rat NRG-1 in samples. No significant cross-reactivity or interference between Rat NRG-1 and analogues was observed.


  • Standard Curve Range: 0,63 to 40 ng/ml
  • Sensitivity: 0,38 ng/ml
  • Inter-Assay: CV% <4,97%
  • Intra-Assay: CV% <4,89%
  • Spike Recovery: 84 to 106%
  • Kit Type: ELISA Kit (Colorimetric)
  • Gene: NRG1

Order Now

SPECIFICATIONS

Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Host
Sample volume 100 µl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR