Optical magnifier
Supplier: OPTIKA
4987
OPTK4987EA 34.2 EUR
OPTK4987
Optical magnifier
Magnifiers
This optical magnifier is designed with front opening and millimetric scale for accurate measurements.
  • Focusing adjustment with 0,1 mm intervals
  • Black linen scale design