Orion™ Storage Sleeve and Base
Supplier: Thermo Orion
810017
ORIO810017EA 46.9 EUR
ORIO810017
Orion™ Storage Sleeve and Base
Electrodes pH Electrodes
Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ electrode storage sleeve and base.
  • It is for use with pH electrodes that are 120 mm or less in length that have a diameter of 12 mm
  • Sleeve protects your electrode from breakage when not in use and will not tip, due to the weighted removable base
  • As an additional convenience, the pH bulb stays conditioned while in storage so that it is ready to use when removed from the sleeve
Limited warranty period: Subject to all warranty requirements and exclusions set out in ourMore Product Information