Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
Supplier: THERMO FISHER DIAGNOSTICS
R4715210
OXOIR4715210EA 1740 EUR
OXOIR4715210
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
Microbiology Test Systems Microorganism Tests
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Balení: Includes 10 preserved organism vials and 10 rehydration vials.