RapID™ System
Supplier: THERMO FISHER DIAGNOSTICS
R8311006 R8325102 R8325106 R8311008 R8309002 R8309003 R8309004 R8311001 R8311002 R8311003 R8311004 R8311007 R8323600 R8311009
OXOIR8311006EA 196 EUR
OXOIR8311006 OXOIR8325102 OXOIR8325106 OXOIR8311008 OXOIR8309002 OXOIR8309003 OXOIR8309004 OXOIR8311001 OXOIR8311002 OXOIR8311003 OXOIR8311004 OXOIR8311007 OXOIR8323600 OXOIR8311009
RapID™ System
Microbiology Test Systems Enzymatic Tests
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions