Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (CF®514)
Supplier: Biotium
P001-514-500 P001-514-125
P001-514-500EA 350 EUR
P001-514-500 P001-514-125
Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (CF®514)
Antibodies
CD4 (clone SK3) is a mouse monoclonal antibody that recognizes the extracellular domain of CD4, a membrane glycoprotein of T lymphocytes. This antibody belongs to the Biotium choice list of select antibodies that have been validated and optimized in-house for optimal performance. The antibody is available conjugated to CF® Dyes and Astral™ tandem dyes. They are supplied in PBS, 0,1% BSA, 0,05% azide.
  • Recommended concentration for flow cytometry: 5 µl per million cells/0,1 ml
  • Protect fluorescent conjugates from light
CD4 interacts with majorMore Product Information