Propidium Iodide solution_SC 1,0 mg/ml water

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide

MQ200

P4864-10ML
P4864-10MLEA 147 EUR
P4864-10ML
Propidium Iodide solution_SC 1,0 mg/ml water
Propidium iodide

Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Suitable for use in fluorescence microscopy, confocal laser scanning microscopy, flow cytometry, and fluorometry. Used as a nuclear counterstain.

Formula: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011921
CAS Number: 25535-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d25c193d13c9581848ebf6ad64efd2c853bada&vid=adcc70f85e90fe9546eb4c24bbf1af34c27f6913&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Color) Faint Orange to Very Dark Orange and Faint Red to Very Dark Red and Faint Purple to Very Dark Purple and Orange-Red and Red-Orange
Appearance (Turbidity) Clear
Appearance (Form), 1.0 mg/mL, H₂O (Solution) Liquid
Purity (HPLC) ≥94%
Extinction Coefficient, 491 to 495 nm in phosphate buffer, pH 7 5400 - 6400
Wavelength 491 - 495 nm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR