Ferrozine mono-sodium salt hydrate, BioXtra, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: FerroZine iron reagent hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate, 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate, PDT disulphonate monosodium salt hydrate

M-Clarity™ quality level = MQ200

P5338-1G P5338-250MG P5338-5G
P5338-1GEA 217 EUR
P5338-1G P5338-250MG P5338-5G
Ferrozine mono-sodium salt hydrate, BioXtra, Sigma-Aldrich®
Ferrozine mono-sodium salt hydrate

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4"-disulfonic acid sodium salt is also known as ferrozine.


  • 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4"-disulfonic acid sodium salt (ABTS) has been used in the ferene-s assay for the accurate determination of intracellular iron from iron oxide nanoparticles
  • It has also been used in ABTS radical scavenging assay to determine the antioxidant activity of honey samples

Formula: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
Melting Pt: ≥300 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00078417
CAS Number: 63451-29-6
EINECS: 274-196-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b37f37e9f5a144e9b4b635ee4e45bcf3d60db43&vid=b5d5cbc9d8bcf6ffc65cc7a802f4a8e125e67211&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Yellow to Yellow-Green
Appearance (Form) Powder
Insoluble matter ≤0.1 %
Chloride Content ≤0.1 %
Aluminum (AL) ≤0.001 %
Phosphorus (P) ≤0.0005 %
Lead (Pb) ≤0.001 %
Iron (Fe) ≤0.0005 %
Copper (Cu) ≤0.0005 %
Zinc (Zn) ≤0.0005 %
Calcium (Ca) ≤0.01 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR