Tablet count kits for water hardness
Supplier: Palintest
AD-1016
PALIAD-1016EA 450 EUR
PALIAD-1016
Tablet count kits for water hardness
Environmental Test Kits
Tablet Count kits provide a simple method of analysis for an important range of water parameters. The tests are particularly useful for the routine control of industrial waters such as boiler waters and cooling water systems, as well as swimming pools, natural waters and similar applications.
  • Cost effective and provides quick results on-site
  • Can be used as a stand alone kit or combined with comparator or photometer
  • Wide range of test concentrations can be measured