qPCR detection of Viral Hemorrhagic Septicemia Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests
Supplier: PRIMERDESIGN
qPCR detection of Viral Hemorrhagic Septicemia Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests