Calibration standard, single element, sodium for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
N9303785
PERCN9303785EA 59.6 EUR
PERCN9303785
Calibration standard, single element, sodium for AAS and ICP-OES, pure grade
Standards Cationic Standards Sodium Standards
Pure Grade standards for AA and ICP-OES.
  • Analyzed by ICP-OES
  • Analyzed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Upozornení: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.