Cleaning cloths, J-Cloth® Plus
Supplier: PGI NONWOVEN
74770 74771 74772 74773 74774 70292
PGIN74770EA 14.4 EUR
PGIN74770 PGIN74771 PGIN74772 PGIN74773 PGIN74774 PGIN70292
Cleaning cloths, J-Cloth® Plus
Wipes Reusable Cleaning Cloths Cloths for General Applications
This reusable cloth is constructed with a unique, open, wavy texture that enables efficient pick up of all types of dirt particles. Designed to provide the ultimate cleaning, wherever hygiene is crucial and HACCP-standards need to be maintained. Ideal for use in food environments.
  • Easy to rinse, fast drying, lightweight: J-Cloth® Plus wipes remain clean and fresh for longer, and can be disposed after use
  • Help to improve efficiency and reduce waste
  • Fully certified to be safe to use in contact with food